Assay Details
Gene Symbol: Tgfb1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191840.1 | NP_001178769.1 | 10 - 11 | 1142 | 92 | |
XM_006230349.2 | XP_006230411.1 | 10 - 11 | 1992 | 92 | ||
XM_006230350.2 | XP_006230412.1 | 10 - 11 | 1114 | 92 | ||
XM_008759920.1 | XP_008758142.1 | 9 - 10 | 2199 | 92 | ||
XM_008759921.1 | XP_008758143.1 | 10 - 11 | 1199 | 92 | ||
GenBank mRNA | AF314960.1 | - | 6 - 7 | 722 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
