Assay Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019347.2 | NP_062220.2 | 8 - 9 | 1236 | 88 | |
NM_177962.3 | NP_808877.2 | 8 - 9 | 1236 | 88 | ||
GenBank mRNA | AF031642.1 | - | 8 - 9 | 1236 | 88 | |
AF041788.1 | - | 8 - 9 | 1236 | 88 | ||
U77971.1 | - | 8 - 9 | 1236 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
