Assay Details
Gene Symbol: Cdc5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053527.1 | NP_445979.1 | 10 - 11 | 1500 | 91 | |
GenBank mRNA | AF000578.2 | - | 10 - 11 | 1500 | 91 | |
BC087007.1 | - | 11 - 12 | 1547 | 91 | ||
BC161839.1 | - | 10 - 11 | 1464 | 9101355 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
