Assay Details
Gene Symbol: Nfkbie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199111.1 | NP_954542.1 | 2 - 3 | 665 | 112 | |
GenBank mRNA | BC061795.1 | - | 2 - 3 | 665 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
