Assay Details
Gene Symbol: Ogfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025708.1 | NP_001020879.1 | 1 - 2 | 354 | 134 | |
XM_008766933.1 | XP_008765155.1 | 1 - 2 | 399 | 134 | ||
GenBank mRNA | BC099205.1 | - | 1 - 2 | 354 | 7384201 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
