Assay Details
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198756.2 | NP_942051.1 | 1 - 2 | 119 | 68 | |
GenBank mRNA | BC058507.1 | - | 1 - 2 | 123 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
