Assay Details
Gene Symbol: Mrps18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198756.2 | NP_942051.1 | 4 - 5 | 378 | 64 | |
GenBank mRNA | BC058507.1 | - | 4 - 5 | 382 | 6889538 | 64 |
FQ215521.1 | - | 5 - 6 | 557 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
