Assay Details
Gene Symbol: Nfya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012865.1 | NP_036997.1 | 4 - 5 | 473 | 108 | |
XM_006244409.2 | XP_006244471.1 | 4 - 5 | 480 | 108 | ||
XM_006244410.1 | XP_006244472.1 | 4 - 5 | 476 | 108 | ||
XM_006244411.2 | XP_006244473.1 | 4 - 5 | 470 | 108 | ||
XM_006244412.2 | XP_006244474.1 | 4 - 5 | 467 | 108 | ||
XM_006244413.1 | XP_006244475.1 | 3 - 4 | 388 | 108 | ||
XM_008766839.1 | XP_008765061.1 | 3 - 4 | 376 | 108 | ||
GenBank mRNA | FQ210330.1 | - | 3 - 4 | 393 | 108 | |
FQ223023.1 | - | 4 - 5 | 485 | 108 | ||
M34238.1 | - | 4 - 5 | 473 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
