Assay Details
Gene Symbol: Frs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017382.1 | NP_001017382.1 | 2 - 3 | 140 | 60 | |
XM_006244436.2 | XP_006244498.1 | 3 - 4 | 657 | 60 | ||
XM_006244437.2 | XP_006244499.1 | 3 - 4 | 511 | 60 | ||
GenBank mRNA | AY972083.1 | - | 2 - 3 | 140 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
