Assay Details
Gene Symbol: Tjap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108203.1 | NP_001101673.1 | 9 - 10 | 857 | 77 | |
XM_006244534.2 | XP_006244596.1 | 8 - 9 | 823 | 77 | ||
XM_006244535.2 | XP_006244597.1 | 6 - 7 | 1118 | 77 | ||
GenBank mRNA | FQ221753.1 | - | 6 - 7 | 1099 | 77 | |
FQ221813.1 | - | 6 - 7 | 1488 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
