Assay Details
Gene Symbol: Mrpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034136.1 | NP_001029308.1 | 1 - 2 | 124 | 104 | |
GenBank mRNA | BC100081.1 | - | 1 - 2 | 124 | 7325700 | 104 |
FQ214924.1 | - | 1 - 2 | 157 | 104 | ||
FQ229593.1 | - | 1 - 2 | 158 | 104 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
