Assay Details
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750668.3 | XP_003750716.3 | 5 - 6 | 715 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
