Assay Details
Gene Symbol: Smap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750667.3 | XP_003750715.3 | 4 - 5 | 628 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
