Assay Details
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008330.1 | NP_001008331.1 | 5 - 6 | 549 | 64 | |
GenBank mRNA | AY325219.1 | - | 3 - 4 | 275 | 64 | |
BC086597.1 | - | 5 - 6 | 549 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
