Assay Details
Gene Symbol: Polr1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008330.1 | NP_001008331.1 | 6 - 7 | 712 | 101 | |
GenBank mRNA | AY325219.1 | - | 4 - 5 | 438 | 101 | |
BC086597.1 | - | 6 - 7 | 712 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
