Assay Details
Gene Symbol: Runx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278483.1 | NP_001265412.1 | 5 - 6 | 913 | 106 | |
XM_006244549.2 | XP_006244611.1 | 4 - 5 | 1114 | 106 | ||
XM_006244550.2 | XP_006244612.1 | 5 - 6 | 1125 | 106 | ||
XM_006244551.2 | XP_006244613.1 | 5 - 6 | 1092 | 106 | ||
GenBank mRNA | AB025797.1 | - | 3 - 4 | 290 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
osteoblast differentiation
endochondral ossification
osteoblast fate commitment
chondrocyte differentiation
chondrocyte development
osteoblast development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
positive regulation of gene expression
T cell differentiation
regulation of ossification
BMP signaling pathway
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
regulation of odontogenesis of dentin-containing tooth
cellular response to fibroblast growth factor stimulus
regulation of osteoblast differentiation
positive regulation of osteoblast differentiation
positive regulation of ossification
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
stem cell differentiation
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
ATP binding
transcription factor binding
protein domain specific binding
histone deacetylase binding
bHLH transcription factor binding
transcription regulatory region DNA binding
repressing transcription factor binding
skeletal system development
osteoblast differentiation
endochondral ossification
osteoblast fate commitment
chondrocyte differentiation
chondrocyte development
osteoblast development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of cell proliferation
positive regulation of gene expression
T cell differentiation
regulation of ossification
BMP signaling pathway
positive regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
regulation of fibroblast growth factor receptor signaling pathway
odontogenesis of dentin-containing tooth
regulation of odontogenesis of dentin-containing tooth
cellular response to fibroblast growth factor stimulus
regulation of osteoblast differentiation
positive regulation of osteoblast differentiation
positive regulation of ossification
negative regulation of smoothened signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
stem cell differentiation
cellular response to BMP stimulus
positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
ATP binding
transcription factor binding
protein domain specific binding
histone deacetylase binding
bHLH transcription factor binding
transcription regulatory region DNA binding
repressing transcription factor binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P051307 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P051312 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P102977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584849 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28846 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051307 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102977 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051312 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051312 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051307 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102977 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P051307 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P051312 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P102977 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16039 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P146816 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P356808 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P259961 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32919 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48635 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46099 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15568 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62703 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10921902 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921900 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921901 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921903 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921904 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10921906 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10921899 | - | - |
Affymetrix Rat Clariom S Assay | PSR0900002460.rn.1 | TC0900000265.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10921899 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365491 | - | - |
