Assay Details
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013131.1 | NP_037263.1 | 3 - 4 | 2370 | 72 | |
GenBank mRNA | M36074.1 | - | 3 - 4 | 2370 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
