Assay Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037784.1 | NP_001032873.1 | 2 - 3 | 357 | 97 | |
XM_006234244.2 | XP_006234306.1 | 2 - 3 | 2571 | 97 | ||
GenBank mRNA | BC098943.1 | - | 2 - 3 | 357 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
