Assay Details
Gene Symbol: Vsig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037784.1 | NP_001032873.1 | 5 - 6 | 834 | 77 | |
XM_006234243.2 | XP_006234305.1 | 6 - 7 | 3157 | 77 | ||
XM_006234244.2 | XP_006234306.1 | 5 - 6 | 3048 | 77 | ||
XM_006234245.2 | XP_006234307.1 | 4 - 5 | 3485 | 77 | ||
XM_006234246.2 | XP_006234308.1 | 4 - 5 | 1736 | 77 | ||
GenBank mRNA | BC098943.1 | - | 5 - 6 | 834 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
