Assay Details
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234737.1 | XP_006234799.1 | 1 - 2 | 90 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006234737.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
