Assay Details
Gene Symbol: Mtch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106488.2 | NP_001099958.2 | 7 - 8 | 550 | 66 | |
GenBank mRNA | BC158687.1 | - | 7 - 8 | 501 | 66 | |
DY311538.1 | - | 501 | 66 | |||
EV779017.1 | - | 571 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
