Assay Details
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037096.1 | NP_001032173.1 | 7 - 8 | 862 | 63 | |
XM_006235066.2 | XP_006235128.1 | 7 - 8 | 1545 | 63 | ||
GenBank mRNA | BC107656.1 | - | 7 - 8 | 862 | 7935449 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
