Assay Details
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107754.2 | NP_001101224.1 | 4 - 5 | 797 | 132 | |
GenBank mRNA | BC168921.1 | - | 4 - 5 | 797 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
