Assay Details
Gene Symbol: Spint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004265.2 | NP_001004265.2 | 8 - 9 | 1494 | 68 | |
XM_006234763.1 | XP_006234825.1 | 9 - 10 | 1506 | 68 | ||
XM_006234764.2 | XP_006234826.1 | 9 - 10 | 1432 | 68 | ||
GenBank mRNA | AB154834.1 | - | 8 - 9 | 1267 | 68 | |
BC097253.1 | - | 8 - 9 | 1494 | 7459422 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
