Assay Details
Gene Symbol: Rbm41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109420.1 | NP_001102890.1 | 6 - 7 | 1130 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P043664 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P087965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692766 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108308 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087965 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043664 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087965 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P043664 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P087965 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P043664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28118 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10939502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939499 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10939503 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10939498 | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00014108.rn.1 | TC0X00002250.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10939498 | - | - |
Rat Genome U34 Set | rc_AI236975_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21541691 | - | - |
