Assay Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107812.2 | NP_001101282.2 | 8 - 9 | 918 | 107 | |
XM_008761908.1 | XP_008760130.1 | 7 - 8 | 763 | 107 | ||
GenBank mRNA | BC167092.1 | - | 8 - 9 | 845 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
