Assay Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107812.2 | NP_001101282.2 | 9 - 10 | 997 | 60 | |
XM_008761908.1 | XP_008760130.1 | 8 - 9 | 842 | 60 | ||
GenBank mRNA | BC167092.1 | - | 9 - 10 | 924 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P536749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746407 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421064 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132901 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22979 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536749 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P536749 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P536749 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P536749 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37290 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38560 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10836839 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836830 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836838 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836834 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836832 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836831 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836837 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836840 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836833 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10836829 | - | - |
Rat Genome 230 2.0 Array | 1375457_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375457_at | - | - |
Affymetrix HT Rat Focus Array | 1375457_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375457_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300006096.rn.1 | TC0300000802.rn.1 | RTA TC |
Rat Expression Set 230 | 1375457_PM_at | - | - |
Rat Expression Set 230 | 1375457_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10836829 | - | - |
Rat Genome U34 Set | rc_AI177300_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22392676 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354287 | - | - |
