Assay Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107812.2 | NP_001101282.2 | 4 - 5 | 558 | 98 | |
XM_008761908.1 | XP_008760130.1 | 3 - 4 | 403 | 98 | ||
GenBank mRNA | BC167092.1 | - | 4 - 5 | 485 | 98 | |
DY575699.1 | - | 3 - 4 | 435 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
