Assay Details
Gene Symbol: Metap1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107812.2 | NP_001101282.2 | 5 - 6 | 601 | 88 | |
XM_008761908.1 | XP_008760130.1 | 4 - 5 | 446 | 88 | ||
GenBank mRNA | BC167092.1 | - | 5 - 6 | 528 | 88 | |
DY575699.1 | - | 4 - 5 | 478 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
