Assay Details
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012184.1 | NP_001012184.1 | 4 - 5 | 835 | 91 | |
XM_006234234.2 | XP_006234296.1 | 4 - 5 | 592 | 91 | ||
XM_006234235.2 | XP_006234297.1 | 4 - 5 | 885 | 91 | ||
XM_006234236.2 | XP_006234298.1 | 4 - 5 | 684 | 91 | ||
XM_006234237.2 | XP_006234299.1 | 4 - 5 | 1444 | 91 | ||
XM_006234238.2 | XP_006234300.1 | 4 - 5 | 592 | 91 | ||
XM_006234239.2 | XP_006234301.1 | 5 - 6 | 1082 | 91 | ||
XM_006234241.2 | XP_006234303.1 | 4 - 5 | 682 | 91 | ||
XM_008761860.1 | XP_008760082.1 | 4 - 5 | 1173 | 91 | ||
GenBank mRNA | BC087094.1 | - | 4 - 5 | 835 | 7191334 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
