Assay Details
Gene Symbol: Gatm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031031.2 | NP_112293.1 | 2 - 3 | 336 | 116 | |
GenBank mRNA | BC081785.1 | - | 2 - 3 | 336 | 116 | |
U07971.1 | - | 2 - 3 | 328 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
