Assay Details
Gene Symbol: Gatm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031031.2 | NP_112293.1 | 7 - 8 | 1101 | 62 | |
GenBank mRNA | BC081785.1 | - | 7 - 8 | 1101 | 62 | |
FQ212973.1 | - | 3 - 4 | 272 | 62 | ||
U07971.1 | - | 7 - 8 | 1093 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
