Assay Details
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106503.1 | NP_001099973.1 | 4 - 5 | 935 | 98 | |
XM_006234917.2 | XP_006234979.1 | 6 - 7 | 1193 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
