Assay Details
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106503.1 | NP_001099973.1 | 5 - 6 | 1016 | 68 | |
XM_006234917.2 | XP_006234979.1 | 7 - 8 | 1274 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P028958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89912 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P028958 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P279041 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P509872 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103108 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10839566 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839577 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839567 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839572 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839576 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839568 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839575 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839573 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839574 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10839565 | - | - |
Rat Genome 230 2.0 Array | 1388635_at | - | - |
Rat Genome 230 2.0 Array | 1388562_at | - | - |
Rat Genome 230 2.0 Array | 1388562_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388635_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373732_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389822_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389822_at | - | - |
Rat Genome 230 2.0 Array | 1373732_at | - | - |
Affymetrix HT Rat Focus Array | 1388562_at | - | - |
Affymetrix HT Rat Focus Array | 1389822_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388562_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1389822_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300011764.rn.1 | TC0300001658.rn.1 | RTA TC |
Rat Expression Set 230 | 1388562_at | - | - |
Rat Expression Set 230 | 1389822_at | - | - |
Rat Expression Set 230 | 1388562_PM_at | - | - |
Rat Expression Set 230 | 1389822_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10839565 | - | - |
Rat Genome U34 Set | rc_AI012979_at | - | - |
Rat Genome U34 Set | rc_AA859179_at | - | - |
Rat Genome U34 Set | rc_AI176601_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21977120 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370199 | - | - |
