Assay Details
Gene Symbol: Qser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139493.1 | NP_001132965.1 | 11 - 12 | 5139 | 157 | |
XM_006234646.2 | XP_006234708.1 | 11 - 12 | 5242 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P539019 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P087419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130254 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P539019 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087419 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P087419 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P539019 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P087419 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P539019 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P539019 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10847951 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847956 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847949 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847943 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847953 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847950 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847948 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847947 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847952 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847946 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847945 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10847955 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10847942 | - | - |
Rat Genome 230 2.0 Array | 1389254_PM_at | - | - |
Rat Genome 230 2.0 Array | 1389254_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300028934.rn.1 | TC0300003793.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300028933.rn.1 | TC0300003793.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300028932.rn.1 | TC0300003793.rn.1 | RTA TC |
Rat Expression Set 230 | 1389254_PM_at | - | - |
Rat Expression Set 230 | 1389254_at | - | - |
Rat Genome U34 Set | rc_AA800519_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21348639 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364101 | - | - |
