Assay Details
Gene Symbol: Qser1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139493.1 | NP_001132965.1 | 5 - 6 | 4315 | 143 | |
XM_006234646.2 | XP_006234708.1 | 5 - 6 | 4418 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
