Assay Details
Gene Symbol: Gyltl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199107.1 | NP_954538.1 | 8 - 9 | 881 | 62 | |
XM_006234562.2 | XP_006234624.1 | 7 - 8 | 2202 | 62 | ||
XM_006234564.2 | XP_006234626.1 | 8 - 9 | 941 | 62 | ||
XM_006234566.2 | XP_006234628.1 | 8 - 9 | 941 | 62 | ||
XM_008762024.1 | XP_008760246.1 | 7 - 8 | 1014 | 62 | ||
XM_008762025.1 | XP_008760247.1 | 7 - 8 | 1129 | 62 | ||
GenBank mRNA | BC061762.1 | - | 8 - 9 | 881 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
