Assay Details
Gene Symbol: Ttl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138536.1 | NP_612545.1 | 4 - 5 | 754 | 86 | |
GenBank mRNA | AF207605.1 | - | 4 - 5 | 754 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
