Assay Details
Gene Symbol: Pdia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017319.1 | NP_059015.1 | 2 - 3 | 326 | 91 | |
GenBank mRNA | BC062393.1 | - | 2 - 3 | 363 | 5621329 | 91 |
D63378.1 | - | 2 - 3 | 326 | 91 | ||
FQ211627.1 | - | 425 | 91 | |||
FQ227046.1 | - | 2 - 3 | 365 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
