Assay Details
Gene Symbol: Katnbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109645.2 | NP_001103115.1 | 1 - 2 | 93 | 148 | |
XM_006234712.2 | XP_006234774.1 | 1 - 2 | 146 | 148 | ||
GenBank mRNA | BC161951.1 | - | 1 - 2 | 93 | 9026012 | 148 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC161951.1, NM_001109645.2, XM_006234712.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
