Assay Details
Gene Symbol: Spi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005892.2 | NP_001005892.1 | 1 - 2 | 196 | 119 | |
GenBank mRNA | AB154364.1 | - | 1 - 2 | 59 | 119 | |
BC091185.1 | - | 1 - 2 | 196 | 7366435 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
