Assay Details
Gene Symbol: Galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013919.1 | NP_001013941.1 | 2 - 3 | 217 | 66 | |
XM_006234904.2 | XP_006234966.1 | 1 - 2 | 672 | 66 | ||
GenBank mRNA | BC083716.1 | - | 2 - 3 | 217 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
