Assay Details
Gene Symbol: Il1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031512.2 | NP_113700.2 | 2 - 3 | 100 | 67 | |
GenBank mRNA | BC091141.1 | - | 2 - 3 | 100 | 7365029 | 67 |
M98820.1 | - | 2 - 3 | 120 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
