Assay Details
Gene Symbol: Rnf128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173349.1 | NP_001166820.1 | 4 - 5 | 1084 | 77 | |
XM_008773419.1 | XP_008771641.1 | 4 - 5 | 1157 | 77 | ||
GenBank mRNA | BC160840.1 | - | 4 - 5 | 945 | 77 | |
BC167061.1 | - | 4 - 5 | 1084 | 77 | ||
FQ210020.1 | - | 4 - 5 | 1072 | 77 | ||
FQ218340.1 | - | 4 - 5 | 1072 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
