Assay Details
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168523.1 | NP_001161995.1 | 10 - 11 | 1073 | 77 | |
GenBank mRNA | FQ220907.1 | - | 10 - 11 | 1046 | 77 | |
FQ229546.1 | - | 10 - 11 | 1067 | 77 | ||
FQ229811.1 | - | 10 - 11 | 1067 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
