Assay Details
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168523.1 | NP_001161995.1 | 4 - 5 | 499 | 84 | |
GenBank mRNA | FQ220907.1 | - | 4 - 5 | 472 | 84 | |
FQ222771.1 | - | 4 - 5 | 491 | 84 | ||
FQ229546.1 | - | 4 - 5 | 492 | 84 | ||
FQ229811.1 | - | 4 - 5 | 493 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
