Assay Details
Gene Symbol: Eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168523.1 | NP_001161995.1 | 7 - 8 | 782 | 130 | |
GenBank mRNA | FQ220907.1 | - | 7 - 8 | 755 | 130 | |
FQ229811.1 | - | 7 - 8 | 776 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
