Assay Details
Gene Symbol: Bcl2l11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234983.2 | XP_006235045.2 | 1 - 2 | 361 | 71 | |
XM_006234985.2 | XP_006235047.2 | 1 - 2 | 361 | 71 | ||
XM_006234987.2 | XP_006235049.2 | 1 - 2 | 361 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
