Assay Details
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762189.1 | XP_008760411.1 | 7 - 8 | 1575 | 96 | |
XM_008762190.1 | XP_008760412.1 | 6 - 7 | 3578 | 96 | ||
GenBank mRNA | FQ212553.1 | - | 3 - 4 | 360 | 96 |
Gene Symbol: Gabpb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135015.1 | NP_001128487.1 | 1 | 883 | 96 | |
XM_006254075.2 | XP_006254137.1 | 2 | 916 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
