Assay Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134854.1 | NP_001128326.1 | 2 - 3 | 740 | 77 | |
XM_006235063.2 | XP_006235125.1 | 3 - 4 | 632 | 77 | ||
XM_006235064.2 | XP_006235126.1 | 3 - 4 | 631 | 77 | ||
XM_006235065.2 | XP_006235127.1 | 3 - 4 | 631 | 77 | ||
XM_008762165.1 | XP_008760387.1 | 3 - 4 | 648 | 77 | ||
GenBank mRNA | BC160836.1 | - | 3 - 4 | 515 | 77 | |
BC161894.1 | - | 2 - 3 | 740 | 77 |
Gene Symbol: RGD1564480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773456.1 | XP_008771678.1 | 1 | 532 | 77 | |
XM_577020.6 | XP_577020.4 | 1 | 531 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
